Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Three-Piece Trubore® Bearing, Ace Glass Incorporated
Supplier: Ace Glass
8042-121 8042-09 8042-117 8042-19 8042-05 8042-135 8042-119 8042-115
89061-764EA 206.25 USD InStock
89061-764 89061-774 89061-762 89061-758 89061-768 89061-756 89061-766 89061-760
Three-Piece Trubore® Bearing, Ace Glass Incorporated
Stirrers Stirring Tools
This piece is a self-aligning three-piece bearing
  • Inner bearing and bushing can be used with any joint size
  • For use with 10mm outer diameter shafts
  • For replacement O-rings, order 89060-984
Complete consists of inner Trubore® glass bearing, nylon bushing with O-ring, and a threaded adapter. Unique design eliminates costly replacement since inner bearing will spin if shaft binds or will self-align at o-ring seal in the event that the motor is slightly cocked. Inner bearing and bushing can be used with any joint size. For use with 10mm outer diameter shafts. For replacement O-rings, order 89060-984.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty