Would you like to log in again?
Trubore® Bearing , Economy, Ace Glass Incorporated
Supplier: Ace Glass
9529-04 9529-10 9529-07
89061-842EA 71.66 USD InStock
89061-842 89075-066 89075-068
Trubore® Bearing , Economy, Ace Glass Incorporated
Stirrers Stirring Tools
Trubore® bearing, heavy duty, standard taper joint, self-aligning
Unique design is forgiving of slight misalignment of stir motor and shaft. Recommended for use with ACE 6 mm precision ground stir shaft.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty