Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Trubore® Bearing, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
8066-55 8066-32 8066-46 8066-28 8066-50 8066-20 8066-33 8066-06 8066-60 8066-43 8066-24
89061-886EA 263.44 USD InStock
89061-886 89061-874 89061-884 89061-872 89061-876 89061-854 89061-848 89097-172 89097-170 89061-882 89061-880
Trubore® Bearing, PTFE, Ace Glass Incorporated
Stirrers Stirring Tools
This self-aligning, lubricant-free PTFE Trubore® glass and PTFE bearing is designed for use with stainless steel or glass 10 mm stirring shafts
  • The compression saddle with O-Ring maintains constant force with little attention
  • Vacuum down to 1 mm
  • Not recommended for precision ground glass shafts
  • Fabricated from borosilicate glass
The lock nut permits stirring in either direction without fear of unthreading.
Delivery information: Available as a complete item, or individual components may be purchased separately. The complete item includes threaded glass body/adapter, 89061-854 PTFE inner bearing, 89061-864 glass-filled PTFE bushing, 89061-868 glass-filled PTFE lock nut, and 89061-870 Rulon compression saddle with O-Ring. Use with glass stirring shafts such as 89061-960 or 89061-970 (among others). For replacement compression saddle size -111 FETFE O-Rings, see 89060-988 series. For size -014 bearing replacement FETFE O-Ring, see 89060-976 series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty