Would you like to log in again?
Stirring Shaft, Stainless Steel, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
8074-04 8074-02 8074-07
89061-962EA 332.03 USD InStock
89061-962 89061-960 89061-964
Stirring Shaft, Stainless Steel, 10 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
Polished stainless steel stirring shafts, 10 mm O.D., drilled type with removable button
When used with a PTFE bearing, a lubricant-free vacuum seal can be realized at speeds up to 200 rpm. Join to stirrer motor using a chuck.
Accessories information: PTFE Gaskets are designed to fit a variety of stirring shafts, such as 89061-938, 89061-946, 89084-572, 89061-962, or 89062-014. Gaskets are also useful as a dust cover, or as a replacement for the gasket supplied with collars (such as 89235-694).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty