Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Heavy Duty Polished Glass Stirrer Shaft, 28 mm, Ace Glass Incorporated
Supplier: Ace Glass
8080-16 8080-14 8080-24 8080-30 8080-12 8080-29 8080-18 8080-25
89062-026EA 1001.59 USD InStock
89062-026 89062-028 89097-176 89097-174 89062-030 76107-028 89062-020 89062-022
Heavy Duty Polished Glass Stirrer Shaft, 28 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
Heavy duty, 28 mm O.D
For Agitators and Turbines Reference the Following Numbers: 89062-152, 89062-174, 89062-176, 71001-576, 71001-578.
glass stirring shaft for use with ACE reactors. Shaft has pre-drilled holes at top and bottom for positive attachment to motor coupling and agitator, respectively. Recommended for use with ACE PTFE Trubore® and mechanical seal bearings. Use with agitators for 28 mm rods. Fits Ace 89054-598 swivel connector and 89054-602 chuck.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty