Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Stirrer Blades, Banana Type, PTFE, 10mm, Ace Glass Incorporated
Supplier: Ace Glass
8087-15 8087-11 8087-13 8087-19 8087-09 8087-05 8087-07
89062-112EA 108.03 USD InStock
89062-112 89062-104 89062-114 89062-106 89062-116 89062-108 89062-110
Stirrer Blades, Banana Type, PTFE, 10mm, Ace Glass Incorporated
Stirrers Stirring Blades
These PTFE, banana shaped stirrer blades are designed for use with 10mm O.D
button-type stir shafts. The 3mm thick blades have a number of perforations, and are designed to closely fit various sizes of round bottom flasks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty