Would you like to log in again?
Single Blade Type Agitator, PTFE, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
8088-10 8088-03
89062-118EA 74.73 USD InStock
89062-118 89062-120
Single Blade Type Agitator, PTFE, 10 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
PTFE agitator with removable blade secured with a PTFE pin
The 76mm (3") long blade (supplied) will fit through a standard taper 29/42 joint. Fits 10mm O.D. drilled type or beaded type shafts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty