Would you like to log in again?
Krytox LVP High Vacuum Grease, Ace Glass Incorporated
Supplier: Ace Glass
8116-10
89062-222EA 284.05 USD InStock
89062-222
Krytox LVP High Vacuum Grease, Ace Glass Incorporated
Lubricants
This grease has very low a vapor pressure, highly inert, and nonflammable
Krytox LVP high vacuum grease is a combination of an extremely low vapor pressure perfluoroalkylpolyether oil and a fluorocarbon resin thickener. This white, buttery grease is designed to lubricate the fittings and accessories of high vacuum systems at operating temperatures down to 10-12 torr at 20°C (1.33 x 10-13 kPa).
The grease is for high-vacuum systems. Superior performance in laboratory and pilot plant equipment, as a lubricant and sealant for stopcocks, valves, fittings and o-rings operating at high vacuum or in hostile environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty