Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Flex-Grip Chuck, Ace Glass Incorporated
Supplier: Ace Glass
8124-26 8124-07 8124-05 8124-12 8124-25 8124-24 8124-13 8124-23 8124-10 8124-30 8124-04 8124-22 8124-20 8124-15 8124-17
89062-261PK 37.46 USD InStock
89062-261 89062-250 89062-252 89062-262 89062-240 89062-254 89062-242 89062-264 89062-256 89062-244 89062-258 89062-236 89062-246 89062-238 89062-248
Flex-Grip Chuck, Ace Glass Incorporated
Stirrers Stirring Tools
Nylon chuck for use with ACE stirring shafts or other shafts of the same dimensions
Chuck has flexible insert which allows for misalignment of shaft without danger of breakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty