Would you like to log in again?
Three-Way Stopcock with Oblique Bore, Ace Glass
Supplier: Ace Glass
8186-04 8186-08
89062-382EA 211.12 USD InStock
89062-382 89062-384
Three-Way Stopcock with Oblique Bore, Ace Glass
Stopcocks
High pressure oblique bore with solid glass plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty