Would you like to log in again?
90° Vacuum Stopcock, PTFE Easy Action Plug, Ace Glass
Supplier: Ace Glass
8193-218 8193-214 8193-04 8193-216
89062-442EA 169.03 USD InStock
89062-442 89062-444 89062-446 89062-440
90° Vacuum Stopcock, PTFE Easy Action Plug, Ace Glass
Stopcocks
Threaded vacuum stopcock with PTFE plug and new hooded handle that permits smooth needle valve adjustment down to 0.1 cc/min flow rate.
Double PTFE ring seals prevent exposure of FETFE backup o-ring to corrosive gases and liquids. Hooded handle can be removed from plug, making plug bakeable. Stem is at 90° angle.
Ordering information: For barrels, see 89062-422. For plugs, see 89062-434. See 89062-496 for replacement o-rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty