Would you like to log in again?
Stopcock Plugs, Straight Bore, 1:5 PTFE, Ace Glass Incorporated
Supplier: Ace Glass
8224-12 8224-08 8224-04 8224-18 8224-16
89062-772EA 39.37 USD InStock
89062-772 89062-774 89062-776 89062-778 89062-770
Stopcock Plugs, Straight Bore, 1:5 PTFE, Ace Glass Incorporated
Stopcocks
Stopcock plug 89062-770 is designed for use with Retainer O-Ring size –011
Stopcock plugs 89062-772, -774, and -776 are designed for use with Retainer O-Ring size –111. Stopcock plug 89062-778 is designed for use with Retainer O-Ring size –208.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty