Would you like to log in again?
High Vacuum Grease, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8230-06
89062-804EA 78.07 USD InStock
89062-804
High Vacuum Grease, Ace Glass Incorporated
Lubricants
Dow Corning High Vacuum Grease
A silicone lubricant that effectively seals and lubricates glass stopcocks, joints and glass-rubber connections. Resistant to most chemicals, heat stable and inert. Supplied in tube, 5.3 oz. (150 g)
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty