Would you like to log in again?
Spherical Joint Stopper Cap, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8274-06 8274-96
89062-890EA 47.91 USD InStock
89062-890 89062-892
Spherical Joint Stopper Cap, Ace Glass Incorporated
Stoppers Glass Stoppers
Cap-type stoppers have spherical joints and are used to seal unused ports on Air Sampling Manifolds
Use pinch clamp to secure caps to ports.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty