Would you like to log in again?
Interchangeable Tissue Grinder, Ace Glass
Supplier: Ace Glass
8345-02 8345-04 8345-10
89063-022EA 149.9 USD InStock
89063-022 89063-020 89063-018
Interchangeable Tissue Grinder, Ace Glass
Homogenizers Tissue Grinders
This is a precision ground instrument for macerating tissue
Rod and barrel are interchangeable. Rod is ground and its entire length and can be reversed for double life. Barrel clearance of .13mm to .2mm meets G.S.A. specifications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty