Would you like to log in again?
Color Stability Tube, Ace Glass
Supplier: Ace Glass
8450-02 8450-10 8450-04
89063-044EA 233.8 USD InStock
89063-044 89063-042 89063-040
Color Stability Tube, Ace Glass
Tubes Centrifuge Tubes
Used in tentative AOCS Method L-15A-58 for measuring the processing color stability of fatty acids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty