Would you like to log in again?
Glass Tubing Connector, 90°, Ace Glass Incorporated
Supplier: Ace Glass
8486-18 8486-12 8486-07 8486-03
89063-132PK 87.37 USD InStock
89063-132 89063-138 89063-136 89063-134
Glass Tubing Connector, 90°, Ace Glass Incorporated
Fittings and Connectors
Each end of this connector features a constriction to hold rubber tubing securely
All connectors feature a 38 mm arm length. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty