Would you like to log in again?
Pressure Tubes with Ace-Thred and Front Seal Plugs, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8648-117 8648-129 8648-123 8648-36 8648-38 8648-245 8648-07 8648-121 8648-48 8648-234 8648-230 8648-136 8648-04 8648-253 8648-53 8648-105 8648-77 8648-34 8648-232 8648-96 8648-308 8648-113 8648-09 8648-55 8648-21 8648-10 8648-43 8648-109 8648-247 8648-249 8648-88 8648-312 8648-25 8648-251 8648-102 8648-115 8648-17 8648-12 8648-127 8648-310
89063-352EA 111.99 USD InStock
89063-352 89063-198 89063-372 89063-174 89063-294 89063-194 89063-172 89063-170 89063-316 76318-586 89063-336 89063-314 89063-214 89063-356 89063-234 89063-354 89063-310 89063-210 89063-298 89063-340 89063-162 89063-382 89063-422 89063-260 89063-380 89063-180 89063-426 89063-408 89063-306 89063-206 89063-302 89063-324 89063-202 89063-300 89063-344 89063-222 89063-166 89063-364 89063-188 89063-320
Pressure Tubes with Ace-Thred and Front Seal Plugs, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are designed for polymerization and analysis of halogens by thermal decomposition in the presence of lime
Larger volume hydrogenation bottles have a flat bottom and a lower recommended operating pressure of 60psi. Bottle pressure ratings are determined at ambient temperature.
Tubes feature Ace-Thred closures which allow for quick and simple plug sealing and removal. Supplied with front seal plug and O-ring for pressure work. This closure system assures a tight and secure seal without the need for any extra equipment. Closures are reusable. Tubes are available in multiple body diameters and lengths, with #7, #15, #25, or #36 Ace-Thred. Tube length is measured below the thread. Tube pressure ratings are determined at 120°C (248°F). Each tube is pressure tested at 1.5 times the rating.
Ordering information: Each tube is supplied with a PTFE front seal plug and FETFE O-ring. Replacement O-rings and O-rings of different composition are available. For monitoring purposes, tube can be assembled with a stainless steel pressure gauge (0–160psi), using an optional adapter. In addition, stainless steel tube extensions are available to allow use of tube in a furnace. Tubes can be fabricated in other lengths and diameters. Tubes equipped with back seal plugs are also available; see 89063-160 series for more information. For replacement tubes and bottles, see 89063-208 series.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty