Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Pressure Tubes with Ace-Thred, Ace Glass
Supplier: Ace Glass
8648-124 8648-46 8648-135 8648-166 8648-23 8648-138 8648-197 8648-193 8648-196 8648-140 8648-192 8648-194 8648-134 8648-27 8648-162 8648-29 8648-191 8648-51 8648-47 8648-165 8648-52 8648-195 8648-157 8648-45 8648-30 8648-33 8648-126 8648-32 8648-120 8648-155 8648-187 8648-133 8648-40 8648-190 8648-42 8648-132 8648-50 8648-26 8648-41 8648-24
89063-330EA 100.54 USD InStock
89063-330 89063-374 89063-296 89063-230 89063-274 76318-590 89063-350 89063-250 89063-370 89063-290 89063-216 89063-238 89063-358 89063-378 89063-334 89063-256 89063-278 89063-332 89063-376 89063-232 89063-254 89063-286 89063-220 89063-362 89063-284 89063-240 89063-360 89063-282 89063-280 89063-208 89063-328 89063-228 89063-348 89063-304 89063-226 89063-368 89063-246 89063-366 89063-288 89063-244
Pressure Tubes with Ace-Thred, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are designed for polymerization and analysis of halogens by thermal decomposition in the presence of lime
They feature the unique Ace-Thred closure system for assurance of a secure seal. Tubes maintain a recommended operating pressure of 150psi. Hydrogenation bottles also feature the Ace-Thred closure system and are flat bottom vessels. These bottles maintain a lower recommended operating pressure, not to exceed 60psi. Tubes and bottles are supplied without plugs and O-rings. Tube length is measured below the thread. Tube pressure ratings are determined at 120°C (248°F). Bottle pressure ratings are determined at ambient temperature. All tubes are pressure tested at 1.5 times the rating.
Ordering information: All pressure tubes and hydrogenation bottles are supplied without closures. These tube-only and bottle-only orders can be used as replacement items for Pressure Tubes with back seal plugs (see 89063-160 series), Pressure Tubes with front seal plugs (see 89063-172 series), or Pressure Tubes with plunger valve (see 89063-386 series).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty