Would you like to log in again?
Diehls-Alder Pressure Tube, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8651-10
89063-438EA 85.44 USD InStock
89063-438
Diehls-Alder Pressure Tube, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy wall with lip for crown cap closure
Used for polymerization and analysis of halogens by thermal decomposition in the presence of lime.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty