Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vacuum Manifold, Walters, Ace Glass Incorporated
Supplier: Ace Glass
8729-25 8729-03 8729-26 8729-40
89063-720EA 571.77 USD InStock
89063-720 89063-722 89063-724 89063-718
Vacuum Manifold, Walters, Ace Glass Incorporated
Manifolds
This greaseless vacuum line is fabricated from Ace-Threds and Easy-Action PTFE stopcocks
Fabricated from borosilicate glass. The top of the main body features a #7 Ace-Thred with a ⅛" NPT nylon adapter for connection to thermocouple vacuum gauge. Each end of the main body has a #25 Ace-Thred for easier cleaning. One end is stoppered with nylon plug, while the other end features a bushing connection to a vacuum trap. The vacuum trap features an Easy-Action, 0 - 10 mm high vacuum stopcock with removable trap. This permits cleaning of the trap without removing vacuum line tubing. The vacuum tubing connection on this trap has a 25 mm O.D. The length of the main body is approximately 61 cm (24"). The vacuum trap is approximately 40.6 cm (16") in length.
Three ports are 0 - 5 mm high vacuum PTFE stopcocks with a #11 Ace-Thred bushing connection to the working vessel. The stopcocks are positioned parallel to the main body to allow operation from front or back. The main body can be reversed for the convenience of left-handed operation.
Ordering information: Available as a complete system, or as individual components. The complete system includes main body, vacuum trap inner tube and body, bushings (89059-226 and 89059-214), stoppers with O-Rings (89052-806 and 89052-828), and #7 Ace-Thred to ?" Swagelok adapter (89052-704).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty