Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Double-Tube Manifold with Threaded Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8729-54 8729-50
89063-726EA 1302.5 USD InStock
89063-726 89063-728
Double-Tube Manifold with Threaded Stopcocks, Ace Glass Incorporated
Manifolds
Greaseless, vacuum/gas manifold features a 0 - 4 mm high vacuum
This manifold has Easy-Action, three-way PTFE stopcocks for takeoffs. The bottom and top manifold tubes each feature a #7 Ace-Thred connection for easy cleaning. The ends are supplied with nylon stopper/plugs with FETFE O-Rings (89052-828). The port connections are 8 mm (1/32") O.D. The vacuum and gas connections are 12.7 mm (1/2") O.D. The distance between the takeoffs is 100 mm (315/16"). Fabricated from borosilicate glass.
Ordering information: Available in three or five port combinations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty