Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vacuum Manifold, Ace Glass Incorporated
Supplier: Ace Glass
8730-27 8730-22 8730-35 8730-29 8730-37 8730-18 8730-16 8730-20 8730-12 8730-33 8730-14 8730-31
89063-750EA 1630.3 USD InStock
89063-750 89063-740 89063-752 89063-730 89063-742 89063-732 89063-744 89063-734 89063-746 89063-736 89063-748 89063-738
Vacuum Manifold, Ace Glass Incorporated
Manifolds
All-glass-and-PTFE vacuum manifold with three, four or five stopcock ports
Each port is a variable opening, 0–10 mm, high vacuum threaded stopcock. Plugs are PTFE with polyethylene or UHDPE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with o-rings as supplied. In the event you need to bake the entire system, order bakeable plugs with PTFE handles. Hose connections on stopcocks and vacuum line are size E for 13-15 mm outer diameter vacuum tubing.
Ordering information: For replacement plugs, see 89062-438 or 89062-580. Available left side connection or right side connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty