Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NMR Tube Tip-Off Manifold, Ace Glass
Supplier: Ace Glass
8731-80 8731-44 8731-34 8731-56 8731-15 8731-19 8731-38 8731-22 8731-12 8731-75 8731-78
89063-760EA 812.17 USD InStock
89063-760 89063-772 89063-762 89063-774 89063-764 89063-754 89063-766 89063-756 89063-768 89063-758 89063-770
NMR Tube Tip-Off Manifold, Ace Glass
Manifolds
Use to tip-off NMR tubes
NMR Tube is inserted in lower Ace-Thred port and tightened via bushing for vacuum-thaw work. Vacuum line is connected to side hose connection. Use with 3/8" or 7/16" inner diameter tubing, size E hose connection. Tube can then be tipped-off with flame. Aluminum shield bonded to nylon bushing helps retard heat transfer. Each port is individually controlled by smooth-acting, semi-needle, threaded PTFE plug to allow removal of one tube while others are still under vacuum, vacuum of 10-6 is common. PTFE plug and tip-off port o-rings are FETFE or Viton. Tip-off ports can be supplied for 5 mm, 8 mm and 10 mm NMR tubes. Manifolds can be supplied with single port or multiple ports of the same size or different sizes.
Delivery information: Complete item consists of glass manifold, PTFE plug(s), bushing(s) with fire shield and O-Rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty