Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vacuum Manifold with Ace-Thred Connections, Ace Glass Incorporated
Supplier: Ace Glass
8734-20 8734-10 8734-14 8734-07
89063-782EA 821.89 USD InStock
89063-782 89063-784 89063-786 89063-780
Vacuum Manifold with Ace-Thred Connections, Ace Glass Incorporated
Manifolds
Glass Manifold with #15 Ace-Thred at either end to allow switching vacuum connection to left or right side
Supplied with three or five 0 - 4 mm high vacuum, Easy-Action stopcocks for sample ports (additional ports can be ordered). Plugs are PTFE with nylon hooded handles and FETFE O-Rings. Ultimate vacuum of 10 - 7 can be realized with standard three o-rings. Overall length of glass manifold is: 9 - 1/8" (232 mm) for three ports; 13 - 5/8" (346 mm) for five ports. Supplied with “Ace-Safe” serrated connector for one end to connect vacuum line
Delivery information: Complete item consists of a glass manifold, 89052-848 end plug with FETFE O-Ring, and Ace- Safe connectors with bushing for 1/4-inch or 1/2-inch tubing connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty