Would you like to log in again?
Vacuum Trap With Serrated Hose Connections, Bent Top Connection, Ace Glass Incorporated
Supplier: Ace Glass
8752-05 8752-17
89063-832EA 213.22 USD InStock
89063-832 89063-914
Vacuum Trap With Serrated Hose Connections, Bent Top Connection, Ace Glass Incorporated
Vacuum Traps
Used for freezing out undesired vapors in the vacuum system and also for use in conjunction with Dewar flasks
Top is bent in line with side tube. Has six-ring hose connections on inlet and outlet tubes. O.D. of smallest ring is 14mm; largest is 16mm. Made of borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty