Would you like to log in again?
Vacuum Trap Assembly, Ace-Thred Top, Ace Glass Incorporated
Supplier: Ace Glass
8748-40
89063-888EA 1907.79 USD InStock
89063-888
Vacuum Trap Assembly, Ace-Thred Top, Ace Glass Incorporated
Vacuum Traps
This twin chambered dry ice vacuum trap is designed to be used with rotary evaporators or any other large solvent evaporation applications
Two ports are located on the sides of chambers; one is for connection to the evaporator, the other to the vacuum source. Both are 10mm O.D., for use with 3/8-inch I.D. tubing. The stopcock at bottom is 4mm PTFE 90° bore, for draining condensate one chamber at a time. Normally, the condensate will collect in the chamber connected nearest to the evaporator until it is filled to crossover between chambers. Then the second chamber will fill. The capacity of the vacuum traps is 450mL each. The capacity of the chambers with the traps inserted is approximately 600mL each. Overall height of the unit is approximately 18 inches.
The unique, squatty design is 18 inches high, and the use of twin chambers offers increased condensing capacity that would be attained only with a much taller, single chambered condenser. The vacuum traps are secured into the main body chambers via #50 Ace-Threds using bushings and o-rings. This type of connection allows for easy removal for cleaning. Fabricated from borosilicate glass.
Ordering information: Complete item includes the twin chamber, two vacuum traps, and two #50 nylon bushings with silicone o-rings. Each item can also be purchased separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty