Would you like to log in again?
Vacuum Trap, Twin Chamber, Ace Glass
Supplier: Ace Glass
8749-20
89063-892EA 624.22 USD InStock
89063-892
Vacuum Trap, Twin Chamber, Ace Glass
Vacuum Traps
Twin-chamber vacuum trap provides double protection against undesired vapors in a vacuum system
Can be used in a 2L Dewar flask. Distance from outside of one body to outside of other is approximately 97mm. Inlet and outlet have serrated fittings with a 14–16mm O.D
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty