Would you like to log in again?
Vacuum Trap, Dewar Type, Ace Glass
Supplier: Ace Glass
8757-10
89063-974EA 469.85 USD InStock
89063-974
Vacuum Trap, Dewar Type, Ace Glass
Vacuum Traps
This Dewar-type vacuum trap has a [ST] 24/40 joint at the bottom
The inside diameter of the coolant section is 50mm, and length is approximately 250mm. The O.D. of the inlet and outlet is 22mm. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty