Would you like to log in again?
Vacuum Traps, Dewar Type with Ace-Thred Inlet/Outlet, Ace Glass
Supplier: Ace Glass
8757-22 8757-24 8757-35
89063-976EA 539.87 USD InStock
89063-976 89063-978 76318-526
Vacuum Traps, Dewar Type with Ace-Thred Inlet/Outlet, Ace Glass
Vacuum Traps
This Dewar type vacuum trap has a [ST] 24/40 inner joint at the bottom
This unit has been fitted with #15 Ace-Threds on the inlet and outlet, for use with Ace-Safe tubing connectors, to make an easy, safe connect/disconnect of tubing. Simply attach 1/2" I.D. tubing to the connector and tighten the bushing 89059-218 to compress the silicone o-ring (supplied with connector) to make a leak-tight seal. Fabricated from borosilicate glass.
Ordering information: Available as a complete item, or components may be purchased separately. Complete item includes glass body, two polypropylene tubing connectors 89052-954 (each with a silicone o-ring), and two #15 Ace-Thred nylon bushings 89059-218.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty