Would you like to log in again?
Dry Ice Trap, Twin Chamber with Stopcock Drain, Ace Glass Incorporated
Supplier: Ace Glass
8758-20 8758-42
89063-980EA 1554.94 USD InStock
89063-980 89063-982
Dry Ice Trap, Twin Chamber with Stopcock Drain, Ace Glass Incorporated
Vacuum Traps
Twin chambered dry ice condenser-trap has dry ice finger condensers in this unit that are NOT removable
Tubing outlets on sides of chambers, for connecting to rotary evaporator and vacuum source, are 10mm O.D. for 3/8-inch I.D. tubing. Condensate stopcock at bottom is 4mm PTFE, 90° bore, that allows draining each chamber individually. Overall height is approximately 13-1/2 inches. Capacity of dry ice finger condensers is 200mL each. Capacity of chambers is approximately 500mL each.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty