Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Single Tube Vacuum Manifold, #15 Ace-Thred Joint Connections and PTFE Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8763-16 8763-46 8763-14 8763-44
89064-010EA 1370.01 USD InStock
89064-010 89064-008 89064-006 89064-004
Single Tube Vacuum Manifold, #15 Ace-Thred Joint Connections and PTFE Stopcocks, Ace Glass Incorporated
Manifolds
Glass manifold with no.15 Ace-Thred at either end to allow switching vacuum connection to left or right side
Supplied with three or five 4 mm straight bore PTFE stopcock sampling ports. Overall length of glass manifold is 230 mm.
Delivery information: Complete item consists of a glass manifold, 89052-848 end plug with FETFE o-ring, and 89052-948 and 89052-954 Ace-Safe connector with bushing for 1/4" or 1/2" tubing connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty