Would you like to log in again?
Septa, Flat, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
8787-43 8787-42 8787-58 8787-40 8787-55 8787-41
89064-076CS 150.62 USD InStock
89064-076 89064-074 89064-082 89064-072 89064-080 89064-078
Septa, Flat, Microscale, Ace Glass Incorporated
Septa
Replacement flat septa are designed for use with microscale reaction vials, or any other V-Bottom reaction vials.
These septa are constructed of white silicone rubber, bonded with a white PTFE facing. They will resist coring when punctured by a syringe needle. All septa are 3 mm thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty