Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Beaded Process Pipe, Extended End Cap, Ace Glass Incorporated

Supplier: Ace Glass
8845-15 8845-11 8845-05 8845-03 8845-09 8845-19 8845-07 8845-17
89064-628EA 205.2 USD InStock
89064-628 89064-618 89064-626 89064-624 89064-622 89064-620 89064-632 89064-630
Beaded Process Pipe, Extended End Cap, Ace Glass Incorporated
Fittings and Connectors
Type of Glass: Borosilicate

Working Pressure:
1/2", 3/4", 1" BP = 100 PSI
1 1/2", 2", 3" BP = 75 PSI
4" = 50 PSI
6" = 30 PSI

Operating Temperature (Maximum):
Freezing point to 450°C
Normal use — 230°C

Glass Thermal Expansion:
32 x 10-7/°C

Glass Therm. Conductivity:
0.73 BTU/ hr. ft.2/F°/ft. at 212°F

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty