Would you like to log in again?
Beaded Process Pipe Short End Cap, Ace Glass Incorporated
Supplier: Ace Glass
8847-02 8847-04 8847-08 8847-10 8847-06
89064-638EA 106.33 USD InStock
89064-638 89064-636 89064-634 89064-642 89064-640
Beaded Process Pipe Short End Cap, Ace Glass Incorporated
Fittings and Connectors
This Beaded Process Pipe Short End Cap is made from Borosilicate glass.
  • The thermal expansion is 32 × 10-7/ °C
  • The shock difference 160 °C
In regards to durability acids, all except Hydrofluoric or Phosphoric with Fluorides. Some durability alkalies attack above 120 °F and above pH11.
Conductivity: 0.73 BTU/ hr. ft.2/F°/ft. at 212 °F
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty