Would you like to log in again?
Beaded Pipe to Conical Pipe Coupling Assembly, Ace Glass Incorporated
Supplier: Ace Glass
8866-03 8866-05 8866-09 8866-07
89064-702EA 932.21 USD InStock
89064-702 89064-700 89064-698 89064-696
Beaded Pipe to Conical Pipe Coupling Assembly, Ace Glass Incorporated
Clamps General Purpose Clamps
To join beaded end pipe to conical end pipe
Ordering information: Kit includes two flanges, inserts, bolts and TFE sheath gasket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty