Would you like to log in again?
Adapter, 75 Degree Angle, Ace Glass Incorporated
Supplier: Ace Glass
5070-15 9052-08 9052-12 5070-10
89051-274EA 86.95 USD InStock
89051-274 89051-318 89064-798 89064-796
Adapter, 75 Degree Angle, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is bent at a 75° angle.
  • [ST] inner joints at both ends
  • Fabricated from borosilicate glass
Adapter has standard taper inner joints at both ends.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty