Would you like to log in again?
Septa, Sleeve Type, Rubber, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9096-254 9096-39 9096-133 9096-32 9096-144 9096-257 9096-249 9096-49 9096-54 9096-57 9096-231 9096-31 9096-243 9096-256 9096-56 9096-25 9096-232 9096-157 9096-43 9096-233 9096-126 9096-239 9096-26 9096-44 9096-131 9096-149 9096-33 9096-156 9096-244 9096-154 9096-143 9096-132 9096-139 9096-226
89064-916PK 121.63 USD InStock
89064-916 89064-938 89064-958 89064-936 89064-914 89064-978 89064-934 89064-956 89064-912 89064-910 89064-954 89064-932 89064-976 89064-974 89064-952 89064-930 89064-950 89064-972 89064-908 89064-906 89064-948 89064-926 89064-904 89064-946 89064-968 89064-924 89064-966 89064-922 89064-964 89064-920 89064-942 89064-940 89064-962 89064-960
Septa, Sleeve Type, Rubber, Ace Glass Incorporated
Stoppers Rubber Stoppers
This sleeve-type septa has a hollow plug
The top is flanged with a sleeve-like extension that folds down over the neck of vessel. The diaphragm can be punctured readily with a syringe needle. Puncture seals automatically after the needle is withdrawn.
Ordering information: Rubber septum are available in white, gray, or red finishes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty