Would you like to log in again?
Adapter, Moisture Trap, Ace Glass Incorporated
Supplier: Ace Glass
5179-07 9101-20
89051-360EA 201.61 USD InStock
89051-360 89064-988
Adapter, Moisture Trap, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a unique adapter used in place of a Dean-Stark moisture test receiver.
  • Top [ST] outer joint
  • Bottom [ST] inner joint
  • Side arm [ST] inner joint
  • Fabricated from borosilicate glass
Simply add a condenser (not included) to the top standard taper outer joint, any graduated funnel from 125mL to 2000mL (not included) to bottom standard taper inner joint, attach a sample flask (not included) to the standard taper inner side arm joint, and you have a moisture test receiver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty