Would you like to log in again?
Gas Inlet Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5265-10 9119-22 9119-02
89051-516EA 93.9 USD InStock
89051-516 89064-992 89064-990
Gas Inlet Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This gas inlet adapter features a side hose connection and standard taper joints.
  • Top [ST] outer joint
  • Bottom [ST] inner joint
  • Side hose connection
  • Fabricated from borosilicate glass
Featuring a side hose connection, with standard taper outer joint at top and standard taper inner joint at bottom. The full-sized outer joints are reinforced.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty