Would you like to log in again?
PIG® Build-A-Berm® Heavy-Duty Barrier Straight Section, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG® Build-A-Berm®
PLR283
89065-410EA 318.24 USD InStock
89065-410
PIG® Build-A-Berm® Heavy-Duty Barrier Straight Section, New Pig
Spill Control Materials
Provides permanent or semi-permanent containment in high-traffic areas on concrete, asphalt or most sealed surfaces
Pre-drilled aluminum mounting strips with steel anchors secure berm to most surfaces; remove anchors to reposition, then seal with caulking (not included).
Create any configuration by combining with other Build-A-Berm components; ends overlap and can be sealed together for liquid-proof containment. When used with other 2"-high Build-A-Berm Barriers, orange color indicates area where drivers can enter and exit.
Constructed of 40-ounce, vinyl-coated fabric with an open-cell foam core; 2"-high berm compresses to approximately 1/4" when driven over; springs back to shape when vehicles pass. Durable enough to withstand indoor or outdoor traffic from trucks, loaded tow motors and other heavy vehicles with sufficient ground clearance.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty