Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GUST® Dusters, Stoner®
Supplier: Stoner
GUST®
94201 94203 94153 94151
89065-924CS 709.15 USD InStock
89065-924 89065-920 89065-922 89065-918
GUST® Dusters, Stoner®
Sprays
GUST® Dusters blast clean, filtered, moisture-free, high-pressue gas to remove dust, dirt, lint, hair, sand, sawdust, moisture, and microscopic debris from hard-to-reach areas or delicate surfaces such as keyboards, printers, computers, cameras, lenses, audio gear, sensitive instruments, and other office equipment or electronics.
  • Blast clean, filtered, moisture-free, high-pressure gas to remove dust, hair, and moisture from hard-to-reach areas or delicate surfaces
  • A unique variable pressure trigger allows for control of dusting pressure
  • Gas is 100% pure, non-flammable, non-ozone depleting, and filtered to 0.2 microns
  • Spray Anyway duster features a 360° valve that allows the user to spray from any angle or upside down without frosting
Duster will not harm glass, optics, metals, plastics, rubber, or hardware. A unique variable pressure trigger allows for control of dusting pressure. Duster gas is 100% pure, non-flammable, non-ozone depleting, and filtered to 0.2 microns.
The Spray Anyway duster features a 360° valve that allows the user to spray from any angle or upside down without frosting.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty