Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Anti-Static Glass Cleaner, Stoner®
Supplier: Stoner
A166
89065-928CS 129.95 USD InStock
89065-928
Anti-Static Glass Cleaner, Stoner®
Cleaning Agents Detergents and Surface Cleansers
Static-dissipating cleaner prevents build-up or static charges when used regularly
Cleaner has a fast-acting, non-abrasive, water-based, foaming formula to remove tough dirt, dust, grease, lint, oil, toner, carbon, fingerprints, and smudges from glass, computer or television screens, CRTs, copiers, scanners, windows, and windshields. It can also be used on plastic, vinyl, and painted surfaces. A special foaming action keeps cleaner in place until wiped, so it will not drip or run. Cleaner dries quickly and leaves no streaks, haze, or film.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty