Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Plastic Surface Cleaner, Stoner®
Supplier: Stoner
A163
89065-930CS 97.2 USD InStock
89065-930
Plastic Surface Cleaner, Stoner®
Cleaning Agents Detergents and Surface Cleansers
Water-based cleaner removes dirt, oil, toner, carbon, fingerprints, fog, and lint from plastic surfaces on computers, printers, photocopiers, telephones, fax machines, and other equipment
It can also be used on glass, vinyl, or painted surfaces. A special foaming action holds the cleaner in place until wiped, so it will not drip or run. Cleaner dries fast, adds shine to surface, and will not leave streaks, haze, or film.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty