Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Multi-Purpose Flux Remover, Stoner®
Supplier: Stoner
94072
89065-932CS 1024.92 USD InStock
89065-932
Multi-Purpose Flux Remover, Stoner®
Soldering Supplies Flux Removers
High-strength cleaner quickly removes rosin-based solder flux to give repairs a factory-new appearance
Remover should not be used on polycarbonate, polystyrene, ABS, display panels, other solvent-sensitive materials or coatings, or on energized equipment. Allow equipment to dry completely before re-energizing. Remover is non-flammable according to ASTM D3065-01 requirements. Supplied in 12oz. aerosol can.
High-pressure eliminates flux and balls remaining from soldering repairs on solvent-resistant components such as connectors, contacts, relays, tuners, servos, switches, solenoids, PCBs, motors, disk drives, and capacitators. Remover evaporates quickly and leaves no residue that could interfere with conformal coating. Low viscosity formula cleans close-tolerance SMT components. Flux remover also functions as contact cleaner and degreaser for PCBs, components, connectors, relays, and boards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty