Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Heavy Duty Electro Klene Cleaner, Stoner®
Supplier: Stoner
Electro Klene
94052
89065-934CS 984.72 USD InStock
89065-934
Heavy Duty Electro Klene Cleaner, Stoner®
Cleaning Agents Detergents and Surface Cleansers
Cleaner loosens and removes heavy oil, grease, dust, dirt, and other residue from electrical and mechanical equipment and precision instruments, such as computers, optics, and industrial controls; it also works on solvent-resistant components such as connectors, contacts, relays, tuners, servos, switches, solenoids, PCBs, motors, disc drives, and capacitators
Cleaner should not be used on polycarbonate, polystyrene, ABS, display panels, or other solvent-sensitive materials or coatings.
Cleaner evaporates quickly and leaves no residue. It is non-flammable and non-conductive, and can be used on some equipment while still in operation. Contains non-CFC cleaning solvent.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty