Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Isopropyl Alcohol Cleaner, Stoner
Supplier: Stoner
D005
89065-938EA 631.89 USD InStock
89065-938
Isopropyl Alcohol Cleaner, Stoner
Cleaning Agents Detergents and Surface Cleansers
Pure isopropyl alcohol acts as a defluxer and general-purpose degreaser to remove dirt, oxides, oils, smoke film, and corrosion from metal and plastic surfaces.
  • IPA is 99.9% pure
  • Evaporates fast
  • Leaves no residue
Apply with squirt bottles, pump spray dispensers, swabs, wipes, or tank dipping techniques.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty