Would you like to log in again?
Dominator Coverall, Attached Hood, MCR Safety
Supplier: MCR Safety
3881XL 3881X2 3881X4 3881X5 3881M 3881L 3881X3
89066-108EA 132.32 USD InStock
89066-108 89066-116 89066-106 89066-110 89066-114 89066-104 89066-112
Dominator Coverall, Attached Hood, MCR Safety
Workwear
The Dominator Series is made tough and roomy for those dirty jobs
It is easily identified by its kelly green color and has additional features that help protect you against splash, grit, and grime. The 3881 is a full coverall with attached hood, is treated to be limited flammability, and is made from a thick .42mm pvc/hi-tensil polyester scrim/pvc material. It features a 30 inch zipper front with snap down double storm flap, has an attached drawstring hood, inner elastic sleeves, and an added 10" x 10" reinforced crotch. Dominate your next job with Dominator series gear.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty