Would you like to log in again?
Light Task Plus 3™ Polyurethane-Coated Cut-Resistant Gloves, Honeywell Safety
Supplier: Honeywell Safety
Light Task Plus 3™
NFD16G/9L NFD16G/10XL NFD16G/8M NFD16G/7S NFD16G/11XXL
89066-228CS 1574.88 USD InStock
89066-228 89066-226 89066-230 89066-224 89066-222
Light Task Plus 3™ Polyurethane-Coated Cut-Resistant Gloves, Honeywell Safety
Gloves
Lightweight, cut level 3 gloves offer the perfect compromise between mechanical protection and wearer comfort.
  • Ideal for material handling and assembly work involving sharp objects
  • Seamless Dyneema® shell is stronger than steel and outperforms aramid fibers in cut and abrasion resistance
  • Tough polyurethane palm coating on palms is abrasion-and tear-resistant yet light enough to ensure dexterity and tactility
  • Glove back is uncoated for ventilation and comfort
  • Knit wrist
Ideal for material handling and assembly work involving sharp objects such as sheet metal, metal shavings, glass, and metal parts.
Certifications: Gloves are tested for cut resistance in accordance with EN 388.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty