Would you like to log in again?
NS4 Series 1/4" Flow Dry Break, Quick Disconnect Coupling, Colder Products Company
Supplier: CPC (Colder Products Company)
NS4D10004 NS4D20006 NS4D22004 NS4D17002 NS4D13006 NS4D22006 NS4D22002 NS4D17004 NS4D24004
89066-504PK 476.51 USD InStock
89066-504 89066-502 89066-492 89066-490 89066-506 89066-496 89066-494 89066-500 89066-498
NS4 Series 1/4" Flow Dry Break, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
1/4" flow quick disconnect couplings offer high flow capacity in a compact size
Materials of construction include glass filled polypropylene, 316 stainless steel and EPDM oring seals.
Engineered with an advanced dry break non spill valve design to eliminate drips and mess enhancing operator and equipment safety. Glass filled polypropylebne for added strength under pressure and color coded for visual identfication.
Ordering information: Accessories not included
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty